Learn More
Medchemexpress LLC Tryptamine-d4 hydrochloride | 340257-60-5 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB2132S15MG
Encompass_Preferred
Tryptamine-d4 hydrochloride is the deuterium labeled Tryptamine hydrochloride. Tryptamine hydrochloride, a monoamine alkaloid, similar to other trace amines, is believed to play a role as a neuromodulator or neurotransmitter.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09911