Learn More
Medchemexpress LLC Tryptamine hydrochloride | 343-94-2 | MFCD00012682 | 100.0% | 196.68 g/mol | C10H13ClN2 | 100 G

Supplier: Medchemexpress LLC HYW014971100G
Tryptamine hydrochloride is the hydrochloride salt of tryptamine supplied as a white to off-white solid for research use. It is commonly used as a biochemical assay reagent and reference standard in studies of indole alkaloids and neurotransmitter-related assays.
- High purity: 99.98%.
- Chemical formula: C10H13ClN2.
- Molecular weight: 196.68 g/mol.
- Appearance: white to off-white solid.
- Storage: solid at 4°C, sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
- Documentation available: data sheet, SDS, COA, LCMS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.