Learn More
Medchemexpress LLC Tryptanthrin | 13220-57-0 | MFCD00012073 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN660750MG
Encompass_Preferred
Tryptanthrin is an indole quinazoline alkaloid that can be found in indigo-bearing plants. It is a potent and orally active cellular Leukotriene (LT) biosynthesis inhibitor and exhibits anticancer activity.
  • Potent and orally active cellular leukotriene (LT) biosynthesis inhibitor
  • Possesses anticancer activity
  • Suppresses expression levels of NOS1, COX-2, and NF-κB
  • Regulates expression levels of IL-2, IL-10, and TNF-α
Catalog No. 50-004-19748