Learn More
Teknova Tryptic Soy Agar, Dry Media.500g
SDP
Supplier:  Teknova T0401
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7344