Learn More
Research Products International Corp Trypto Soy Broth Liquid 1 L
SDP
Supplier:  Research Products International Corp T501501000.0
Encompass_Preferred
Trypto Soy Broth (TSB) Liquid is a nutrient-rich medium commonly used for the cultivation and growth of various microorganisms, particularly bacteria. It contains a blend of tryptone, soy peptone, and other nutrients, providing a suitable environment for the proliferation of a wide range of bacteria. 
TSB liquid is often employed in microbiology laboratories for tasks such as bacterial growth studies, antibiotic susceptibility testing, and microbial fermentation experiments. Its versatility and ability to support robust microbial growth make it a staple in microbiological research.
Catalog No. 50-000-52423