Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptone, Spectrum™ Chemical
SDP

Specifications

Chemical Name or Material Tryptone
Color Powder
Physical Form Solid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-529
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
T1333100GM
100 g Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Specifications

Specifications

Tryptone
Powder
Solid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.