Learn More
MOLTOX Molecular Toxicology Inc Tryptone Yeast Broth, 55 mL/bottle, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 26517
Encompass_Preferred
This Tryptone Yeast Broth consists of Peptone and Yeast extract only. Does not contain the addition of NaCl. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-740