Learn More
Medchemexpress LLC Tryptophan-1-13C | 81201-92-5 | 205.22 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0623S3
Encompass_Preferred
L-Tryptophan-1-13C is the 13C-labeled L-Tryptophan, an essential amino acid and precursor of serotonin, melatonin, and vitamin B3. This compound is intended for research use only.
  • Purity of 99.94%
  • Solid, white to off-white appearance
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-90329