Learn More
Quanterix Inc TSA Cyanine 5 Reagent Pack 50-150 Slides
SDP
Supplier:  Quanterix Inc SAT705A001EA
Encompass
TSA Cyanine 3 (Cy3) reagent pack, for amplification of signal in immunohistochemistry (IHC), immunofluorescence (IF) or in situ hybridization protocols. These reagents amplify by depositing extra Cy5 in the localized area of the probe or antibody.
Catalog No. NC1611910