Learn More
Aviva Systems Biology Corp TSEN2 Antibody - C-terminal region (ARP62409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSEN2. It was validated on Western Blot. . Alternate names: MGC2776, MGC4440, PCH2B, SEN2, SEN2LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-287-675