Learn More
Genscript Corporation TSH CA8 mAb Mouse
Supplier:  Genscript Corporation V08102100
Encompass
Thyroid stimulating hormone (TSH) is a pituitary hormone that stimulates the thyroid gland to secrete thyroxine (T4) and triiodothyronine (T3) It is a glycoprotein consisting of the alpha and the beta subunit TSH serves as a sensitive and specific biomarker for evaluating thyroid function It is a useful marker for monitoring patients under substitution therapy
Catalog No. 50-000-59194
Only null left
Add to Cart
Request a Quote