Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 21.5X300
Supplier:  Tosoh Bioscience LLC 0014460
Encompass
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 10 micrometer; Pore Size: 80 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-356
Only null left
Add to Cart
Request a Quote