Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 4.6X250
Supplier:  Tosoh Bioscience LLC 0013071
Encompass
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 4.6mm I.D. x 25cm Length; Stainless Steel
Catalog No. 50-851-302