Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST Q 10X100
Supplier:  Tosoh Bioscience LLC 0021411
Encompass
Column, Ion Exchange Chromatography; Cation Exchange; Tosoh TSKgel BioAssist S; Media: Polymer; Particle: 13 micrometer; 10mm I.D. x 10cm Length; PEEK
Catalog No. 50-851-675
Only null left
Add to Cart
Request a Quote