Learn More
Tosoh Bioscience LLC TSKGEL CM-5PW 7.5X75
Supplier:  Tosoh Bioscience LLC 0013068
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel CM-5PW; Matrix: hydroxylated methacrylic polymer; Stainless-steel; Counter ion: Nasup+ Particle: 10u m; Pore size: 1000angstrom pH: 2.0 -12; 7.5mm I.D. x 7.5cm
Catalog No. 50-851-300