Learn More
Tosoh Bioscience LLC TSKGEL CN-80TS 4.6X150
Supplier:  Tosoh Bioscience LLC 0017348
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel CN-80Ts; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 4.6mm I.D. x 15cm Length; Stainless Steel - Standard
Catalog No. 50-851-435