Learn More
Tosoh Bioscience LLC TSKGEL G2000SWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0008540
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G2000SWxl; Media: Silica; Particle: 5 micrometer; Pore Size: 125 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-268