Learn More
Tosoh Bioscience LLC TSKGEL G3000SW GL 8X300
Supplier:  Tosoh Bioscience LLC 0008800
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G3000SW Glass; Particle: 10 micrometer; Pore Size: 250 angstrom; Media: Silica; 8mm I.D. x 30cm Length; Glass
Catalog No. 50-851-284