Learn More
Tosoh Bioscience LLC TSKGEL G5000PW 7.5X300
Supplier:  Tosoh Bioscience LLC 0005764
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G5000PW; Media: Polymer; Particle: 17 micrometer; Pore Size: 1000 angstrom; 7.5mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-133