Learn More
Tosoh Bioscience LLC TSKGEL GMPWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0008025
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel GMPWxl; Media: Mixed bed polymer; Particle: 13 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-246