Learn More
Tosoh Bioscience LLC TSKGEL OCTADECYL-4PW 21.5X150
Supplier:  Tosoh Bioscience LLC 0016257
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Octadecyl-4PW; Media: Polymer; Particle: 13 micrometer; Pore Size: 500 angstrom; 21.5mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-413
Only null left
Add to Cart
Request a Quote