Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 2X50 5UM
Supplier:  Tosoh Bioscience LLC 0021457
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;2mm x 5cm Length; Stainless Steel
Catalog No. 50-851-682