Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS QA 2X250
Supplier:  Tosoh Bioscience LLC 0018769
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Ts QA; Media: Silica; Particle: 5 micrometer; 2mm x 25cm Length; Stainless Steel
Catalog No. 50-851-523