Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW RP 21.5X150
Supplier:  Tosoh Bioscience LLC 0016260
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Phenyl-5PW RP; Media: Polymer; Particle: 13 micrometer; Pore Size: 1000 angstrom; 21.5mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-416
Add to Cart
Request a Quote