Learn More
Tosoh Bioscience LLC TSKGEL Q-STAT
Supplier: Tosoh Bioscience LLC 0021961
Column, Anion Exchange Chromatography; Tosoh TSKgel Q-STAT; Media: Polymer; Particle: 7 micrometer; Pore Size: non-porous; 4.6mm x 10cm Length; Stainless Steel