Learn More
Tosoh Bioscience LLC TSKGEL SUPER-PHENYL 2X50 2UM
Supplier:  Tosoh Bioscience LLC 0020017
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Super-Phenyl; Media: Silica; Particle: 2 micrometer; Pore Size: 110 angstrom; 2mm I.D. x 5cm Length; Stainless Steel
Catalog No. 50-851-621