Learn More
Tosoh Bioscience LLC TSKGEL SUPERHZ1000 6X150 5UM
Supplier:  Tosoh Bioscience LLC 0019302
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel SuperHZ1000; Media: Polymer; Particle: 3 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-536