Learn More
IW Tremont Co Inc LabExact Filter Binderless glass microfiber TSS 1.5um 4.25cm dia. Use for EPA Methods 2540C and 2540D for testing dissolved and suspended solids in water and wastewater, gravimetric analysis of air pollutants 100/pk
SDP
Supplier:  IW Tremont Co Inc LETSS4250
Encompass_Preferred
LabExact Grade TSS is a Binderless high efficiency (HEPA type) filter medium with particle retention of 1.50um. Liquid filtration properties have been specifically designed for EPA Methods 2540C and 2540D for testing dissolved and suspended solids in water and wastewater. High flow rate with high capacity. Binderless borosilicate glass fiber media has no added extractables to aid in the elimination of sample contamination. Excellent wet strength. Other common applications include gravimetric analysis of air pollutants, membrane support pads, membrane prefilters, clarification of reagent and buffer solutions, filtration of eluent for HPLC and moisture analysis pads. ¿Suspended solids are those that can be retained on a water filter and are capable of settling out of the water column onto the stream bottom when stream velocities are low. They include silt, clay, plankton, organic wastes, and inorganic precipitates such as those from acid mine drainage.¿
Catalog No. 50-223-7214