Learn More
Cayman Chemical TStosTRon ELISA ntIsRum 500dtn
SDP
Supplier:  Cayman Chemical 482702500 DTN
Encompass_Preferred
An ACSL4 inhibitor (IC50 0.38 UM) selective for ACSL4 over ACSL3 (IC50 >50 UM) and PPARY (IC50 >10 UM)
Catalog No. 50-289-2372