Learn More
Medchemexpress LLC TSTU | 105832-38-0 | MFCD00077875 | ≥99.0% | 301.05 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01141250G
Encompass_Preferred
TSTU is a coupling reagent used in peptide chemistry. It converts carboxylic acids into N-hydroxysuccinimidyl (NHS) esters and can be applied to attach labels to peptides, proteins, and similar molecules. It is also suitable for the in situ formation of succinimide esters in mixed aqueous/organic media for amide bond formation reactions.
  • Converts carboxylic acids into N-hydroxysuccinimidyl (NHS) esters.
  • Suitable for attaching labels to peptides and proteins.
  • Applied for in situ formation of succinimide esters.
  • Used in amide bond formation reactions.
Catalog No. 50-004-54989