Learn More
SAKURA FINETEK USA INC TT MICROTOME BLADE 500/CS
Supplier:  SAKURA FINETEK USA INC 7089
Encompass
Accu-Edge Disposable Microtome Blades Blade angle: 22", Tissue-Tek Feather Low Profile Blades; 500/case. 0.24 (T) x 8 (H) x 80 (L) mm
Catalog No. NC3449537