Learn More
Medchemexpress LLC TTA-A2 | 953778-63-7 | 99.3% | 378.39 | 100 MG
SDP
Catalog No. 5000438317
Encompass_Preferred
TTA-A2 is a potent, selective, and orally active T-type voltage-gated calcium channel antagonist with reduced pregnane X receptor (PXR) activation. It is equally potent against the Cav3.1 (a1G) and Cav3.2 (a1H) channels with IC50 values of 89 nM and 92 nM, respectively, at -80 and -100 mV holding potentials. TTA-A2 can be used for the research of various human neurological diseases, including sleep disorders and epilepsy.
  • Potent, selective, orally active T-type voltage-gated calcium channel antagonist.
  • Reduced pregnane X receptor (PXR) activation.
  • Equally potent against Cav3.1 and Cav3.2 channels.
  • Used in research for neurological diseases, sleep disorders, and epilepsy.
  • Exhibits high affinity in α1I binding assay (Ki of 1.2 nM).
  • Excellent selectivity over hERG potassium and L-type calcium channels.
  • Modifies sleep architecture in rats and mice.
Catalog No. 50-004-38317 Supplier Medchemexpress LLC Supplier No. HY111828100MG
TTA-A2 is a potent, selective, and orally active T-type voltage-gated calcium channel antagonist with reduced pregnane X receptor (PXR) activation. It is equally potent against the Cav3.1 (a1G) and Cav3.2 (a1H) channels with IC50 values of 89 nM and 92 nM, respectively, at -80 and -100 mV holding potentials. TTA-A2 can be used for the research of various human neurological diseases, including sleep disorders and epilepsy.
  • Potent, selective, orally active T-type voltage-gated calcium channel antagonist.
  • Reduced pregnane X receptor (PXR) activation.
  • Equally potent against Cav3.1 and Cav3.2 channels.
  • Used in research for neurological diseases, sleep disorders, and epilepsy.
  • Exhibits high affinity in α1I binding assay (Ki of 1.2 nM).
  • Excellent selectivity over hERG potassium and L-type calcium channels.
  • Modifies sleep architecture in rats and mice.