Learn More
Medchemexpress LLC TTHA | 869-52-3 | 98.0% | 494.45 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW013718
Encompass_Preferred
TTHA is a complexonate that exhibits high affinity for light rare earth elements (LREE), particularly for La3+ in ternary mixtures and Y3+ in quinary mixtures. It is intended for research use only.
  • High affinity for light rare earth elements (LREE)
  • Specifically effective for La3+ in ternary mixtures
  • Specifically effective for Y3+ in quinary mixtures
  • Intended for research use only
Catalog No. 50-003-66763