Learn More
Cayman Chemical ttramthylrhodamIn IsoTHOc 10mg
SDP
Supplier:  Cayman Chemical 1959310 MG
Encompass_Preferred
An amine-reactive derivative of rhodamine that is used as a fluorescent label for antibodies and other probes (ex/em max = 552/575 nm)
Catalog No. 50-298-3441