Learn More
Bioworld TT - SDS Running Buffer (Cathode) (10X), 1 L
Supplier:  Bioworld 214200292
Encompass_Preferred
Product DescriptionTris-Tricine (TT)-SDS running buffer has a 10X concentration and a pH of 8.3. It contains 1M Tris-base, 1M Tricine and 1% SDS.The tricine system requires SDS in sample and running buffers for best results. Tricine-SDS PAGE is commonly used to separate proteins that range in molecular weight between 1 – 100 kDa but is preferred for the resolution of proteins smaller than 30 kDa.
Catalog No. 50-198-662