Learn More
Azenta US Inc TU,0.9,EXT,2CD,CP HBRK,TUL96RK
Supplier:  Azenta US Inc 68100112
Encompass
0.9ml Dual-coded Tube, 96-format, External Thread, 2D Code and Human Readable Number on base, capped, 10 racks per case, HighBase rack, with TubeLock.AZENTA US INC. mfg# 68-1001-12
Catalog No. NC2298093