Learn More
AVANTOR MASTERFLEX (VWR LLC) C-Flex Tubing Braided; 1/2""id, 3/4""OD, 1/8"" Wall, 15'
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0643307
Encompass
Applications: For use with cell cultures, vessel and tank transfer, pump applications and cold storage. Characteristics: C-Flex formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. Certifications: Complies with USP Class VI standards. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: Autoclavable and gamma sterilizable.
Catalog No. NC1813950