Learn More
*SAINT GOBAIN PERF PLASTICS BEAVERTON Tygon&trade, Sanitary Silicone Tubing Formula 3350, I.D., 1/4 in. (6.4mm), Wall Thickness, 3/32 in. (2.4mm), O.D., 7/16 in. (11mm)
Supplier:  *SAINT GOBAIN PERF PLASTICS BEAVERTON ABW00018
Encompass_Preferred
Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications. Ultrasmooth inner bore reduces risk of particle entrapment, inhibits microscopic bacterial buildup during fluid transfer, and allows thorough cleaning and sterilization.Tasteless, odorless, Nonpyrogenic, nontoxic, nonhemolytic. Low extractable content helps to maintain integrity and purity of transported fluidsSmooth inner surface provides low adsorption for improved fluid flow (hydrophobic surface also improves fluid flow)Maximum recommended operating temperature 200°C (392°F)Autoclavable; also can be radiation- and gas-sterilizedDurometer hardness: Shore A, 50. Not intended for use as an implant material.
Catalog No. 50-206-7722