Learn More
Saint Gobain Performance Plastics Corp Chemfluor™, Fluoropolymer Tubing, FEP, I.D., 0.0625 in., O.D., 0.125 in.
Supplier:  Saint Gobain Performance Plastics Corp TSFE14012503150
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals. Contains no additives or plasticizersInert to virtually all chemicals. Nonprotein binding.
Catalog No. 50-206-7716