Learn More
Saint Gobain Performance Plastics Corp Chemfluor&trade, Fluoropolymer Tubing, FEP, I.D., 0.156 in., O.D., 0.25 in.
Supplier:  Saint Gobain Performance Plastics Corp TSFE14025004750
Encompass_Preferred
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals. Contains no additives or plasticizersInert to virtually all chemicals. Nonprotein binding.
Catalog No. 50-206-7717