Learn More
Saint Gobain Performance Plastics TUB TYG 2375 1/16 x 3/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics AJK00003
Encompass_Preferred
Saint-Gobain Tygon Ultra-Chemical-Resistant Tubing Formula 2375 Diameter Inner 0.063 in. 1.6 mm Diameter Outer 3/16 in. 4.8 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15 m
Catalog No. 50-206-9022