Learn More
Saint Gobain Performance Plastics TUB TYG 2375 1/4 x 3/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics AJK00017
Encompass_Preferred
Saint-Gobain Tygon Ultra-Chemical-Resistant Tubing Formula 2375 Diameter Inner 1/4 in. 6.4 mm Diameter Outer 3/8 in. 9.5 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15 m
Catalog No. 50-206-9025