Learn More
Saint Gobain Performance Plastics TUB TYG 2375 1/8 x 1/4 50FT/PK
Supplier:  Saint Gobain Performance Plastics AJK00007
Encompass_Preferred
Saint-Gobain Tygon Ultra-Chemical-Resistant Tubing Formula 2375 Diameter Inner 0.13 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15 m
Catalog No. 50-206-9023