Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Thermoplastic, 1-1/4 in ID, 50 ft
Supplier:  Accuflow Systems ACF00070
Encompass_Preferred
Tygon E-3603 is a clear, flexible thermoplastic tubing formulated for medical and diagnostic fluid handling applications that require visual inspection, chemical compatibility, and low extractables. It provides consistent dimensional tolerance and a smooth inner bore to minimize buildup and carryover in fluid transfer systems.
  • Clear thermoplastic construction for visual fluid inspection.
  • 1-1/4 in inner diameter, 1-5/8 in outer diameter, 3/16 in wall.
  • 50 ft continuous length for system installation.
  • Phthalate-free and BPA-free formulation.
  • Biocompatible and non-contaminating; low extractables.
  • Smooth inner surface reduces carryover and eases cleaning.
  • High lot-to-lot consistency for repeatable performance.
  • Suitable for IVD analyzers, medical instrumentation, and laboratory fluid transfer.
  • Compliant with REACH and California Proposition 65.
Catalog No. NC3238174