Learn More
Aviva Systems Biology Corp TUBA4A Antibody - middle region (ARP40179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBA4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TUBA4A. Alternate names: FLJ30169, H2-ALPHA, TUBA1ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53178