Learn More
TARGETMOL CHEMICALS INC TUBACIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T63275MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubacin is a highly potent and selective reversible cell-permeable HDAC6 inhibitor with an IC50 of 4 nM approximately 350-fold selectivity over HDAC1. purity: 98%
Catalog No. 50-257-9430