Learn More
Selleck Chemical LLC Tubacin 5mg 537049-40-4
SDP
Supplier:  Selleck Chemical LLC S22395MG
Encompass_Preferred
Tubacin is a highly potent and selective, reversible, cell-permeable HDAC6 inhibitor with an IC50 of 4 nM in a cell-free assay, approximately 350-fold selectivity over HDAC1. Tubacin reduces the replication of the Japanese Encephalitis Virus via the decrease of viral RNA synthesis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0769176