Learn More
Selleck Chemical LLC Tubastatin A
SDP
Supplier:  Selleck Chemical LLC S8049200MG
Encompass_Preferred
Tubastatin A is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay It is selective against all the other isozymes (1000-fold) except HDAC8 (57-fold) Tubastatin A promotes autophagy and increases apoptosis
Catalog No. 50-000-00217