Learn More
Selleck Chemical LLC Tubastatin A HCl S2627-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S262710MM/1ML
Encompass_Preferred
Tubastatin A HCl is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay It is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more)
Catalog No. 50-311-2373