Learn More
Ace Glass, Inc. Tube, pressure, 120mL, 150psi, 38.1mm OD, 17.8cm long, complete with #15 back-seal plug
SDP
Supplier:  Ace Glass, Inc. 864889
Encompass_Preferred
100mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 38.1mm O.D. x 17.8cm long.
Catalog No. NC0362913